Learn More
Eurofins Discoverx PathHunter Human CFTR-ΔF508 MEM-EA Pharmacotrafficking Cell-Based Assay Kit (U2OS)
Catalog No. 5000455366
Encompass
The PathHunter Human CFTR-ΔF508 MEM-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant CFTR's trafficking to the membrane. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55366 Supplier Eurofins Discoverx Supplier No. 7930987C3
The PathHunter Human CFTR-ΔF508 MEM-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant CFTR's trafficking to the membrane. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.