Learn More
Eurofins Discoverx PathHunter Human EGFR/EGFR Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455271
Encompass
The PathHunter Human EGFR/EGFR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of EGFR/EGFR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55271 Supplier Eurofins Discoverx Supplier No. 7930989C3
The PathHunter Human EGFR/EGFR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of EGFR/EGFR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.