Learn More
Eurofins Discoverx PathHunter Mouse RHO(P23H) ENDO-EA Pharmacotrafficking Cell-Based Assay Kit (U2OS)
Catalog No. 5000455290
Encompass
The PathHunter Mouse RHO(P23H) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant RHO's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55290 Supplier Eurofins Discoverx Supplier No. 7930990C3
The PathHunter Mouse RHO(P23H) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant RHO's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.