Learn More
Eurofins Discoverx PathHunter Human AVPR2 (S167T) ENDO-EA Pharmacotrafficking Cell-Based Assay Kit (U2OS)
Catalog No. 5000455291
Encompass
The PathHunter Human AVPR2 (S167T) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant AVPR2's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55291 Supplier Eurofins Discoverx Supplier No. 7930992C3
The PathHunter Human AVPR2 (S167T) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant AVPR2's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.