Learn More
Eurofins Discoverx PathHunter Human IL-29 (IFNRL1/IL10RB) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455294
Encompass
The PathHunter Human IL-29 (IFNRL1/IL10RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IFNRL1/IL10RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55294 Supplier Eurofins Discoverx Supplier No. 7930993C3
The PathHunter Human IL-29 (IFNRL1/IL10RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IFNRL1/IL10RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.