Learn More
Eurofins Discoverx PathHunter Human RANK/IkB Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455310
Encompass
The PathHunter Human RANK IkB Stable Cell Line Functional Kinase Assay (U2OS) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55310 Supplier Eurofins Discoverx Supplier No. 7930994C3
The PathHunter Human RANK IkB Stable Cell Line Functional Kinase Assay (U2OS) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.