Learn More
Eurofins Discoverx PathHunter Human VEGF (KDR/KDR) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455350
Encompass
The PathHunter Human VEGF (KDR/KDR) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of KDR/KDR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55350 Supplier Eurofins Discoverx Supplier No. 7930996C1
The PathHunter Human VEGF (KDR/KDR) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of KDR/KDR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.