Learn More
Eurofins Discoverx PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455292
Encompass
The PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL2RB/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55292 Supplier Eurofins Discoverx Supplier No. 7930998C3
The PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL2RB/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.