Learn More
Eurofins Discoverx PathHunter Human IL-7 (IL7R/IL2RG) Dimerization Cell-Based Assay Kit (DLD1)
Catalog No. 5000455348
Encompass
The PathHunter Human IL-7 (IL7R/IL2RG) Stable Cell Line Dimerization Assay (DLD1) measures ligand-induced dimerization of IL7R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55348 Supplier Eurofins Discoverx Supplier No. 7930997C13
The PathHunter Human IL-7 (IL7R/IL2RG) Stable Cell Line Dimerization Assay (DLD1) measures ligand-induced dimerization of IL7R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.