Learn More
Eurofins Discoverx PathHunter Human IL-13 (IL4R/IL13RA1) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455269
Encompass
The PathHunter Human IL-13 (IL4R/IL13RA1) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL4R/IL13RA1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55269 Supplier Eurofins Discoverx Supplier No. 7931001C1
The PathHunter Human IL-13 (IL4R/IL13RA1) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL4R/IL13RA1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.