Learn More
Eurofins Discoverx PathHunter Human IL-31 (OSMRb/IL31R) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455272
Encompass
The PathHunter Human IL-31 (OSMRb/IL31R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of OSMRb/IL31R. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55272 Supplier Eurofins Discoverx Supplier No. 7931002C3
The PathHunter Human IL-31 (OSMRb/IL31R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of OSMRb/IL31R. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.