Learn More
Eurofins Discoverx PathHunter Human MC4R(T162I) ENDO-EA Pharmacotrafficking Cell-Based Assay Kit (U2OS)
Catalog No. 5000455233
Encompass
The PathHunter Human MC4R(T162I) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant MC4R's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55233 Supplier Eurofins Discoverx Supplier No. 7931004C3
Add to Cart
Add to Cart
The PathHunter Human MC4R(T162I) ENDO-EA Pharmacotrafficking Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant MC4R's trafficking to early endosomes. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.