Learn More
Eurofins Discoverx PathHunter Human IL-2 (IL-2R (IL2RA/IL2RB/IL2RG)) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455346
Encompass
The PathHunter Human IL-2 (IL-2R (IL2RA/IL2RB/IL2RG)) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL2RB/IL2RG/IL2RA. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55346 Supplier Eurofins Discoverx Supplier No. 7931003C3
The PathHunter Human IL-2 (IL-2R (IL2RA/IL2RB/IL2RG)) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL2RB/IL2RG/IL2RA. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.