Learn More
Eurofins Discoverx PathHunter Human IL-23 (IL23R/IL12RB1) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455286
Encompass
The PathHunter Human IL-23 (IL23R/IL12RB1) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL23R/IL12RB1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55286 Supplier Eurofins Discoverx Supplier No. 7931007C3
The PathHunter Human IL-23 (IL23R/IL12RB1) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL23R/IL12RB1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.