Learn More
Eurofins Discoverx PathHunter Human BMP-6 (BMPR1B/ACVR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455166
Encompass
The PathHunter Human BMP-6 (BMPR1B/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55166 Supplier Eurofins Discoverx Supplier No. 7931010C3
Add to Cart
Add to Cart
The PathHunter Human BMP-6 (BMPR1B/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.