Learn More
Eurofins Discoverx PathHunter Human TGFBR1/ACVR2 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455273
Encompass
The PathHunter Human TGFBR1/ACVR2 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of TGFBR1A/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55273 Supplier Eurofins Discoverx Supplier No. 7931012C3
The PathHunter Human TGFBR1/ACVR2 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of TGFBR1A/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.