Learn More
Eurofins Discoverx PathHunter Human MC5R β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000456015
Encompass
The PathHunter Human MC5R β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure MC5R (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56015 Supplier Eurofins Discoverx Supplier No. 7930231C3
The PathHunter Human MC5R β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure MC5R (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.