Learn More
Eurofins Discoverx PathHunter Mouse Insulin Receptor B (INSRb) Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455371
Encompass
The PathHunter Mouse Insulin Receptor B (INSRb) Stable Cell Line Functional Kinase Assay (U2OS) measures INSRb receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55371 Supplier Eurofins Discoverx Supplier No. 7931017C3
The PathHunter Mouse Insulin Receptor B (INSRb) Stable Cell Line Functional Kinase Assay (U2OS) measures INSRb receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.