Learn More
Eurofins Discoverx PathHunter Human TSLP (IL7R/CRLF2) Dimerization Cell-Based Assay Kit (DLD1)
Catalog No. 5000455212
Encompass
The PathHunter Human TSLP (IL7R/CRLF2) Stable Cell Line Dimerization Assay (DLD1) measures ligand-induced dimerization of IL7R/CRLF2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55212 Supplier Eurofins Discoverx Supplier No. 7931019C13
The PathHunter Human TSLP (IL7R/CRLF2) Stable Cell Line Dimerization Assay (DLD1) measures ligand-induced dimerization of IL7R/CRLF2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.