Learn More
Eurofins Discoverx PathHunter Human Tie2 Functional Kinase Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455168
Encompass
The PathHunter Human Tie2 Stable Cell Line Functional Kinase Assay (HEK 293) measures Tie2 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55168 Supplier Eurofins Discoverx Supplier No. 7931024C1
The PathHunter Human Tie2 Stable Cell Line Functional Kinase Assay (HEK 293) measures Tie2 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.