Learn More
Eurofins Discoverx PathHunter Human IL-22 (IL22RA1/IL10RB) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455433
Encompass
The PathHunter Human IL-22 (IL22RA1/IL10RB) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL22RA/IL10RB1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55433 Supplier Eurofins Discoverx Supplier No. 7931026C1
The PathHunter Human IL-22 (IL22RA1/IL10RB) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL22RA/IL10RB1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.