Learn More
Eurofins Discoverx PathHunter Human IL-24 (IL20RA/IL20RB) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455434
Encompass
The PathHunter Human IL-24 (IL20RA/IL20RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL20R1/IL20R2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55434 Supplier Eurofins Discoverx Supplier No. 7931027C3
The PathHunter Human IL-24 (IL20RA/IL20RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL20R1/IL20R2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.