Learn More
Eurofins Discoverx PathHunter Human IL-11 (IL11RA/IL6ST) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455208
Encompass
The PathHunter Human IL-11 (IL11RA/IL6ST) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL11R/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55208 Supplier Eurofins Discoverx Supplier No. 7931028C1
The PathHunter Human IL-11 (IL11RA/IL6ST) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL11R/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.