Learn More
Eurofins Discoverx PathHunter Human IL-21 (IL21R/IL2RG) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455210
Encompass
The PathHunter Human IL-21 (IL21R/IL2RG) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL21R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55210 Supplier Eurofins Discoverx Supplier No. 7931035C3
The PathHunter Human IL-21 (IL21R/IL2RG) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL21R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.