Learn More
Eurofins Discoverx PathHunter Human IL-1 alpha (IL1R1/IL1RAP) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455351
Encompass
The PathHunter Human IL-1 alpha (IL1R1/IL1RAP) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL1R1/IL1RAP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55351 Supplier Eurofins Discoverx Supplier No. 7931032C3
The PathHunter Human IL-1 alpha (IL1R1/IL1RAP) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL1R1/IL1RAP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.