Learn More
Eurofins Discoverx PathHunter Human BMP-4 (BMPR1A/ACVR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455360
Encompass
The PathHunter Human BMP-4 (BMPR1A/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1A/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55360 Supplier Eurofins Discoverx Supplier No. 7931037C3
The PathHunter Human BMP-4 (BMPR1A/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1A/ACVR2A. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.