Learn More
Eurofins Discoverx PathHunter Human IL-9 (IL9R/IL2RG) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455435
Encompass
The PathHunter Human IL-9 (IL9R/IL2RG) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL9R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55435 Supplier Eurofins Discoverx Supplier No. 7931036C1
The PathHunter Human IL-9 (IL9R/IL2RG) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL9R/IL2RG. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.