Learn More
Eurofins Discoverx PathHunter Human Neuregulin-1 beta 2 (ErbB2/ErbB3) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455221
Encompass
The PathHunter Human Neuregulin-1 beta 2 (ErbB2/ErbB3) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB2/ErbB3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55221 Supplier Eurofins Discoverx Supplier No. 7931042C3
The PathHunter Human Neuregulin-1 beta 2 (ErbB2/ErbB3) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB2/ErbB3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.