Learn More
Eurofins Discoverx PathHunter Human IL-6 (IL6RA/IL6ST) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455299
Encompass
The PathHunter Human IL-6 (IL6RA/IL6ST) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL6RA/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55299 Supplier Eurofins Discoverx Supplier No. 7931045C3
Add to Cart
Add to Cart
The PathHunter Human IL-6 (IL6RA/IL6ST) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL6RA/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.