Learn More
Eurofins Discoverx PathHunter Human GDF-11 (TGFBR1/ACVR2B) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455438
Encompass
The PathHunter Human GDF-11 (TGFBR1/ACVR2B) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of TGFBR1/ACVR2B. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55438 Supplier Eurofins Discoverx Supplier No. 7931046C3
The PathHunter Human GDF-11 (TGFBR1/ACVR2B) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of TGFBR1/ACVR2B. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.