Learn More
Eurofins Discoverx PathHunter Human BMP-6 (BMPR1B/ACVR2B) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455288
Encompass
The PathHunter Human BMP-6 (BMPR1B/ACVR2B) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/ACVR2B. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55288 Supplier Eurofins Discoverx Supplier No. 7931052C3
The PathHunter Human BMP-6 (BMPR1B/ACVR2B) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/ACVR2B. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.