Learn More
Eurofins Discoverx PathHunter Human BMP-6 (BMPR1B/BMPR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455171
Encompass
The PathHunter Human BMP-6 (BMPR1B/BMPR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/BMPR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55171 Supplier Eurofins Discoverx Supplier No. 7931053C3
Add to Cart
Add to Cart
The PathHunter Human BMP-6 (BMPR1B/BMPR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of BMPR1B/BMPR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.