Learn More
Eurofins Discoverx PathHunter Human c-Ret-GF Ralpha 1 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455367
Encompass
The PathHunter Human c-Ret-GFR alpha 1 Stable Cell Line Functional Kinase Assay (U2OS) measures c-Ret-GFRα1 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55367 Supplier Eurofins Discoverx Supplier No. 7931054C3
The PathHunter Human c-Ret-GFR alpha 1 Stable Cell Line Functional Kinase Assay (U2OS) measures c-Ret-GFRα1 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.