Learn More
Eurofins Discoverx PathHunter Human FGFR3/FGFR3 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455312
Encompass
The PathHunter Human FGFR3/FGFR3 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of FGFR3/FGFR3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55312 Supplier Eurofins Discoverx Supplier No. 7931055C3
The PathHunter Human FGFR3/FGFR3 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of FGFR3/FGFR3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.