Learn More
Eurofins Discoverx PathHunter Human NIK Signaling Cell-Based Assay Kit (U2OS)
Catalog No. 5000455222
Encompass
The PathHunter Human NIK Signaling Stable Cell Line Assay (U2OS) measures NFκB Signaling by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55222 Supplier Eurofins Discoverx Supplier No. 7931059C3
The PathHunter Human NIK Signaling Stable Cell Line Assay (U2OS) measures NFκB Signaling by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.