Learn More
Eurofins Discoverx PathHunter Human Oncostatin M (OSMR/IL6ST) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455237
Encompass
The PathHunter Human Oncostatin M (OSMR/IL6ST) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of OSMR/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55237 Supplier Eurofins Discoverx Supplier No. 7931056C3
The PathHunter Human Oncostatin M (OSMR/IL6ST) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of OSMR/IL6ST. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.