Learn More
Eurofins Discoverx PathHunter Human FGFR4-beta-Klotho Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455223
Encompass
The PathHunter Human FGFR4-beta-Klotho Stable Cell Line Functional Kinase Assay (U2OS) measures FGFR4-β-Klotho receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55223 Supplier Eurofins Discoverx Supplier No. 7931060C3
The PathHunter Human FGFR4-beta-Klotho Stable Cell Line Functional Kinase Assay (U2OS) measures FGFR4-β-Klotho receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.