Learn More
Eurofins Discoverx PathHunter Human M-CSF (CSF1R/CSF1R) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455369
Encompass
The PathHunter Human M-CSF (CSF1R/CSF1R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of CSF1R/CSF1R. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55369 Supplier Eurofins Discoverx Supplier No. 7931061C3
Add to Cart
Add to Cart
The PathHunter Human M-CSF (CSF1R/CSF1R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of CSF1R/CSF1R. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.