Learn More
Eurofins Discoverx PathHunter Human KCNH2(G601S) MEM-EA Pharmacotrafficking Cell-Based Assay Kit (U2OS)
Catalog No. 5000455287
Encompass
The PathHunter Human KCNH2(G601S) MEM-EA Pharmacotrafficking Stable Cell Line Assay Kit (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant KCNH2's trafficking to the membrane. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55287 Supplier Eurofins Discoverx Supplier No. 7931064C3
The PathHunter Human KCNH2(G601S) MEM-EA Pharmacotrafficking Stable Cell Line Assay Kit (U2OS) contains a stable clonal cell line used to measure pharmacochaperone-based rescue of the mutant KCNH2's trafficking to the membrane. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.