Learn More
Eurofins Discoverx PathHunter Human TNF alpha (IL17RD/TNFR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455437
Encompass
The PathHunter Human TNF alpha (IL17RD/TNFR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL17RD/TNFR2 . This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55437 Supplier Eurofins Discoverx Supplier No. 7931068C3
The PathHunter Human TNF alpha (IL17RD/TNFR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL17RD/TNFR2 . This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.