Learn More
Eurofins Discoverx PathHunter Human Activin A (ACVR1B/BMPR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455368
Encompass
The PathHunter Human Activin A (ACVR1B/BMPR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVR1B/BMPR2 . This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55368 Supplier Eurofins Discoverx Supplier No. 7931072C3
The PathHunter Human Activin A (ACVR1B/BMPR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVR1B/BMPR2 . This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.