Learn More
Eurofins Discoverx PathHunter Human GLP-1 (GLP-1 Receptor) Total GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455170
Encompass
The PathHunter Human GLP-1 (GLP1R) Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GLP1R (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55170 Supplier Eurofins Discoverx Supplier No. 7931075C3
The PathHunter Human GLP-1 (GLP1R) Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GLP1R (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.