Learn More
Eurofins Discoverx PathHunter Human VISTA/VISTA Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455224
Encompass
The PathHunter Human VISTA/VISTA Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of the two target proteins. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55224 Supplier Eurofins Discoverx Supplier No. 7931079C3
The PathHunter Human VISTA/VISTA Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of the two target proteins. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.