Learn More
Eurofins Discoverx PathHunter Human TIM3/TIM3 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455440
Encompass
The PathHunter Human TIM3/TIM3 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of the two target proteins. eXpress kits contain all assay materials: cells, reagents, and plates. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55440 Supplier Eurofins Discoverx Supplier No. 7931082C3
The PathHunter Human TIM3/TIM3 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of the two target proteins. eXpress kits contain all assay materials: cells, reagents, and plates. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.