Learn More
Eurofins Discoverx PathHunter Human OX40 Signaling Cell-Based Assay Kit (U2OS)
Catalog No. 5000455441
Encompass
The PathHunter Human OX40 (NIK) Signaling Stable Cell Line Assay (U2OS) measures OX40 signal by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55441 Supplier Eurofins Discoverx Supplier No. 7931080C3
Add to Cart
Add to Cart
The PathHunter Human OX40 (NIK) Signaling Stable Cell Line Assay (U2OS) measures OX40 signal by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.