Learn More
Eurofins Discoverx PathHunter Human 4-1BB (CD137) Signaling Cell-Based Assay Kit (U2OS)
Catalog No. 5000455370
Encompass
The PathHunter Human 4-1BB (CD137) (NIK) Signaling Stable Cell Line Assay (U2OS) measures CD137 signal by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55370 Supplier Eurofins Discoverx Supplier No. 7931089C3
The PathHunter Human 4-1BB (CD137) (NIK) Signaling Stable Cell Line Assay (U2OS) measures CD137 signal by activation-induced stabilization of NIK. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.