Learn More
Eurofins Discoverx PathHunter Human CTLA4 Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455226
Encompass
The PathHunter Human CTLA4 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures CTLA4 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55226 Supplier Eurofins Discoverx Supplier No. 7931094C19
The PathHunter Human CTLA4 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures CTLA4 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.