Learn More
Eurofins Discoverx PathHunter Human FGFR3(G380R)/FGFR3(G380R) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455362
Encompass
The PathHunter Human FGFR3(G380R)/FGFR3(G380R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of FGFR3(G380R)/FGFR3(G380R). G380R version. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55362 Supplier Eurofins Discoverx Supplier No. 7931091C3
The PathHunter Human FGFR3(G380R)/FGFR3(G380R) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of FGFR3(G380R)/FGFR3(G380R). G380R version. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.