Learn More
Eurofins Discoverx PathHunter Human IFN beta (IFNAR1/IFNAR2) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455301
Encompass
The PathHunter Human IFN beta (IFNAR1/IFNAR2) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IFNAR1/IFNAR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55301 Supplier Eurofins Discoverx Supplier No. 7931097C1
The PathHunter Human IFN beta (IFNAR1/IFNAR2) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IFNAR1/IFNAR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.