Learn More
Eurofins Discoverx PathHunter Human IFN gamma (IFNGR1/IFNGR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455850
Encompass
The PathHunter Human IFN gamma (IFNGR1/IFNGR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IFNGR1/IFNGR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55850 Supplier Eurofins Discoverx Supplier No. 7931098C3
The PathHunter Human IFN gamma (IFNGR1/IFNGR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IFNGR1/IFNGR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.